Fmoc-​Val-​Cit-​PAB 95%

Supplier: AFG BIOSCIENCE LLC

C56014-1G C56014-2G C56014-500MG
76620-206EA 155.76 CAD
76620-206 76620-208 76620-204
Fmoc-​Val-​Cit-​PAB 95%
Fmoc-​Val-​Cit-​PAB
Formula: C₃₃H₃₉N₅O₆
MW: 601.7 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22417106
CAS Number: 159858-22-7

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR