tert-Butyl N-[(4-Aminophenyl)Methyl]-N-Methylcarbamate ≥95%
Supplier: AFG Bioscience
C56012-1G
C56012-500MG
C56012-5G
76620-200EA
778.81
CAD
76620-200
76620-198
76620-202
tert-Butyl N-[(4-Aminophenyl)Methyl]-N-Methylcarbamate ≥95%
tert-Butyl N-[(4-Aminophenyl)Methyl]-N-Methylcarbamate
Formula:
C₁₃H₂₀N₂O₂ MW: 236.31 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12413374 CAS Number: 225240-83-5 |
Specification Test Results
Purity Limit | > 95% |
Appearance | Orange Solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...