Diglycine-Biotin 95%

Supplier: AFG Bioscience

C55753-10MG C55753-5MG
76620-196EA 712.09 CAD
76620-196 76620-194
Diglycine-Biotin 95%
Diglycine-Biotin
Formula: C₁₆H₂₈N₆O₄S
Storage Temperature: Freezer
CAS Number: 1363159-69-6

Order Now

Specification Test Results

Purity Limit 95%
Appearance white solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR