3-​[2-​(2-​methoxyethoxy)​ethoxy]​prop-​1-​yne 95%

Supplier: AFG Bioscience

Synonyms: mPEG2-Propyne

C55289-1G C55289-2G C55289-500MG
76620-190EA 316.47 CAD
76620-190 76620-192 76620-188
3-​[2-​(2-​methoxyethoxy)​ethoxy]​prop-​1-​yne 95%
3-​[2-​(2-​methoxyethoxy)​ethoxy]​prop-​1-​yne
Formula: C₈H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD30531705
CAS Number: 89635-79-0

Order Now

Specification Test Results

Purity Limit 95%
Appearance Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR