UDP-rhamnose 95%

Supplier: AFG Bioscience

C55203-10MG C55203-5MG
76620-186EA 1022.13 CAD
76620-186 76620-184
UDP-rhamnose 95%
UDP-rhamnose
Formula: C₁₅H₂₄N₂O₁₆P₂
Storage Temperature: Freezer
CAS Number: 1955-26-6

Order Now

Specification Test Results

Purity Limit 95%
Appearance White solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR