3-Amino-3-deoxy-D-ribose, HCl salt ≥95%

Supplier: AFG BIOSCIENCE LLC

C54767-100MG C54767-250MG C54767-50MG
76620-148EA 467.29 CAD
76620-148 76620-150 76620-146
3-Amino-3-deoxy-D-ribose, HCl salt ≥95%
3-Amino-3-deoxy-D-ribose, HCl salt
Formula: C₅ClH₁₂NO₄
MW: 185.61 g/mol
Storage Temperature: Refrigerator
CAS Number: 20590-58-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR