2-Acetamido-1,3,4-tri-O-acetyl-2-deoxy-α-L-fucopyranose ≥95%

Supplier: AFG Bioscience

Synonyms: α-L-FucNAc triacetate, 2-Acetamido-1,3,4-tri-O-acetyl-2,6-dideoxy-α-L-galactopyranose

C54762-10MG C54762-25MG C54762-5MG
76620-142EA 389.41 CAD
76620-142 76620-144 76620-140
2-Acetamido-1,3,4-tri-O-acetyl-2-deoxy-α-L-fucopyranose ≥95%
2-Acetamido-1,3,4-tri-O-acetyl-2-deoxy-α-L-fucopyranose
Formula: C₁₄H₂₁NO₈
Storage Temperature: Refrigerator
CAS Number: 115509-87-0

Order Now

Specification Test Results

Purity Limit > 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR