Benzyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-galactopyranoside ≥97%

Supplier: AFG Bioscience

C54719-1G C54719-2G C54719-5G
76620-128EA 237.36 CAD
76620-128 76620-130 76620-132
Benzyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-galactopyranoside ≥97%
Benzyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-galactopyranoside
Formula: C₂₁H₂₇NO₉
Storage Temperature: Refrigerator
CAS Number: 41355-94-6

Order Now

Specification Test Results

Purity Limit > 97%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR