4-Nitrophenyl β-D-lactopyranoside heptaacetate 95%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
4-Nitrophenyl 2,3,6-tri-O-acetyl-4-O-(2,3,4,6-tetra-O-acetyl-β-D-galactopyranosyl)-β-D-glucopyranoside
C54705-1G
C54705-2G
C54705-500MG
76620-124EA
1027.33
CAD
76620-124
76620-126
76620-122
4-Nitrophenyl β-D-lactopyranoside heptaacetate 95%
4-Nitrophenyl β-D-lactopyranoside heptaacetate
Formula:
C₃₂H₃₉NO₂₀ MW: 757.65 g/mol Storage Temperature: Freezer |
CAS Number:
84034-75-3 |
Specification Test Results
Purity Limit | 95% |
Appearance | White solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...