1-Amino-1-deoxy-β-D-galactose 95%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
β-D-Galactosylamine, 1-Amino-1-deoxy-β-D-galactose
C54351-1G
C54351-250MG
C54351-500MG
76620-074EA
632.96
CAD
76620-074
76620-070
76620-072
1-Amino-1-deoxy-β-D-galactose 95%
1-Amino-1-deoxy-β-D-galactose
Formula:
C₆H₁₃NO₅ MW: 179.17 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00047541 CAS Number: 74867-91-7 |
Specification Test Results
Purity Limit | 95% |
Appearance | White powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...