2-Amino-2-deoxy-L-fucose HCl salt ≥95%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
L-Fucosamine HCl salt, 2-Amino-2,6-dideoxy-L-galactose HCl salt
C54323-10MG
C54323-25MG
C54323-5MG
76620-066EA
630.48
CAD
76620-066
76620-068
76620-064
2-Amino-2-deoxy-L-fucose HCl salt ≥95%
2-Amino-2-deoxy-L-fucose HCl salt
Formula:
C₆ClH₁₄NO₄ MW: 199.63 g/mol Storage Temperature: Refrigerator |
CAS Number:
7577-62-0 |
Specification Test Results
Purity Limit | > 95% |
Appearance | white powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...