Allyl α-​D-​mannopyranoside 95%

Supplier: AFG Bioscience

C54285-1G C54285-2G C54285-500MG
76620-060EA 395.6 CAD
76620-060 76620-062 76620-058
Allyl α-​D-​mannopyranoside 95%
Allyl α-​D-​mannopyranoside
Formula: C₉H₁₆O₆
Storage Temperature: Refrigerator
MDL Number: MFCD08274515
CAS Number: 41308-76-3

Order Now

Specification Test Results

Purity Limit 95%
Appearance White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR