2-Acetamido-2-deoxy-D-talopyranose ≥95%

Supplier: AFG Bioscience

Synonyms: N-Acetyl-D-talosamine, TalNAc

C54122-10MG C54122-2MG C54122-5MG
76620-038EA 712.09 CAD
76620-038 76620-034 76620-036
2-Acetamido-2-deoxy-D-talopyranose ≥95%
2-Acetamido-2-deoxy-D-talopyranose
Formula: C₈H₁₅NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD08703994
CAS Number: 282727-46-2

Order Now

Specification Test Results

Purity Limit > 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR