Methyl 2-​acetamido-​2-​deoxy-β-​D-​galactopyranoside 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: Methyl β-N-acetyl-D-galactosamine

C54051-1G C54051-250MG C54051-500MG
76620-026EA 934.59 CAD
76620-026 76620-022 76620-024
Methyl 2-​acetamido-​2-​deoxy-β-​D-​galactopyranoside 95%
Methyl 2-​acetamido-​2-​deoxy-β-​D-​galactopyranoside
Formula: C₉H₁₇NO₆
MW: 235.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077377
CAS Number: 22256-76-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR