Methyl 2-​acetamido-​2-​deoxy-​β-​D-​glucopyranoside 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: Methyl N-acetyl-β-D-glucosamine

C54048-1G C54048-5G
76620-006EA 316.47 CAD
76620-006 76620-008
Methyl 2-​acetamido-​2-​deoxy-​β-​D-​glucopyranoside 95%
Methyl 2-​acetamido-​2-​deoxy-​β-​D-​glucopyranoside
Formula: C₉H₁₇NO₆
MW: 235.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00069801
CAS Number: 3946-01-8

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR