4-Nitrophenyl-β-D-lactopyranoside 98%
Supplier: AFG Bioscience
Synonyms:
Gal1-β-4Glc1-β-PNP
C53889-500MG
C53889-1G
C53889-2G
C53889-100MG
76619-990EA
395.6
CAD
76619-990
76619-992
76619-994
77888-956
4-Nitrophenyl-β-D-lactopyranoside 98%
4-Nitrophenyl-β-D-lactopyranoside
Formula:
C₁₈H₂₅NO₁₃ MW: 463.39 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00069843 CAS Number: 4419-94-7 |
Specification Test Results
Purity Limit | 98% |
Appearance | White solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...