N-Acetyl-3-O-β-D-galactopyranosyl-β-D-galactosamine methyl glycoside ≥95%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
Galb1-3GalNAcb1-OMe, b-Galactosyl 1-3 N-acetyl galactosamine methyl glycoside, Methyl galacto-N-biose
C53877-10MG
C53877-25MG
C53877-5MG
76619-986EA
553.84
CAD
76619-986
76619-988
76619-984
N-Acetyl-3-O-β-D-galactopyranosyl-β-D-galactosamine methyl glycoside ≥95%
N-Acetyl-3-O-β-D-galactopyranosyl-β-D-galactosamine methyl glycoside
Formula:
C₁₅H₂₇NO₁₁ MW: 397.37 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01862835 CAS Number: 88274-25-3 |
Specification Test Results
Purity Limit | > 95% |
Appearance | White Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...