4-Aminophenyl β-D-glucopyranoside 95%

Supplier: AFG Bioscience

C53750-1G C53750-500MG C53750-250MG
76619-964EA 467.29 CAD
76619-964 76619-962 77888-935
4-Aminophenyl β-D-glucopyranoside 95%
4-Aminophenyl β-D-glucopyranoside
Formula: C₁₂H₁₇NO₆
Storage Temperature: Freezer
MDL Number: MFCD00067365
CAS Number: 20818-25-1

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR