Lewis-a trisaccharide ≥95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: Lea-Trisaccharide, Galactosyl-β-1,3-(fucosyl-α-1,4)-N-acetylglucosamine, α-L-Fuc-(1→4)-[β-D-Gal-(1→3)]-D-GlcNAc

C53684-10MG C53684-25MG
76619-946EA 949.44 CAD
76619-946 76619-948
Lewis-a trisaccharide ≥95%
Lewis-a trisaccharide
Formula: C₂₀H₃₅NO₁₅
MW: 529.49 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00214078
CAS Number: 56570-03-7

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR