2-​O-​Methyl-​β-​D-​N-​acetylneuraminic Acid ≥95%

Supplier: AFG Bioscience

C53620-10MG C53620-25MG C53620-50MG
76619-930EA 395.6 CAD
76619-930 76619-932 76619-934
2-​O-​Methyl-​β-​D-​N-​acetylneuraminic Acid ≥95%
2-​O-​Methyl-​β-​D-​N-​acetylneuraminic Acid
Formula: C₁₂H₂₁NO₉
Storage Temperature: Freezer
MDL Number: MFCD00084809
CAS Number: 23755-35-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White To Off-White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR