Chloro 2,3,5-tri-O-p-chlorobenzoyl-α-D-ribofuranoside ≥95%

Supplier: AFG Bioscience

C53527-100MG C53527-250MG C53527-500MG
76619-924EA 316.47 CAD
76619-924 76619-926 76619-928
Chloro 2,3,5-tri-O-p-chlorobenzoyl-α-D-ribofuranoside ≥95%
Chloro 2,3,5-tri-O-p-chlorobenzoyl-α-D-ribofuranoside
Formula: C₂₆H₁₈Cl₄O₇
Storage Temperature: Freezer
CAS Number: 125598-74-5

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White To Off-White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR