2,3,4-Tri-O-benzyl-L-fucopyranose ≥95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 6-Deoxy-2,3,4-tri-O-(phenylmethyl)-L-galactose

C53352-1G C53352-5G
76619-908EA 237.36 CAD
76619-908 76619-910
2,3,4-Tri-O-benzyl-L-fucopyranose ≥95%
2,3,4-Tri-O-benzyl-L-fucopyranose
Formula: C₂₇H₃₀O₅
MW: 434.52 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00153596
CAS Number: 60431-34-7

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR