1,​2,​3,​6-​tetra-​O-​acetyl-​β-​D-​glucopyranose ≥95%

Supplier: AFG Bioscience

C53339-1G C53339-250MG C53339-500MG
76619-906EA 1557.64 CAD
76619-906 76619-902 76619-904
1,​2,​3,​6-​tetra-​O-​acetyl-​β-​D-​glucopyranose ≥95%
1,​2,​3,​6-​tetra-​O-​acetyl-​β-​D-​glucopyranose
Formula: C₁₄H₂₀O₁₀
Storage Temperature: Refrigerator
CAS Number: 27086-15-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR