Phenyl 2,3,4,6-tetra-O-benzyl-a-D-thiomannopyranoside 98%

Supplier: AFG Bioscience

C53266-1G C53266-5G
76619-872EA 316.47 CAD
76619-872 76619-874
Phenyl 2,3,4,6-tetra-O-benzyl-a-D-thiomannopyranoside 98%
Phenyl 2,3,4,6-tetra-O-benzyl-a-D-thiomannopyranoside
Formula: C₄₀H₄₀O₅S
Storage Temperature: Refrigerator
MDL Number: MFCD06657858
CAS Number: 116501-53-2

Order Now

Specification Test Results

Purity Limit 98%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR