Methyl 2,3,4-triacetate-α-D-glucopyranoside 95%

Supplier: AFG Bioscience

C53248-1G C53248-2G C53248-5G
76619-866EA 474.73 CAD
76619-866 76619-868 76619-870
Methyl 2,3,4-triacetate-α-D-glucopyranoside 95%
Methyl 2,3,4-triacetate-α-D-glucopyranoside
Formula: C₁₃H₂₀O₉
Storage Temperature: Freezer
MDL Number: MFCD11113467
CAS Number: 7432-72-6

Order Now

Specification Test Results

Purity Limit 95%
Appearance White solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR