Methyl-2,3,4-tri-O-benzyl-ɑ-D-glucopyranoside ≥95%

Supplier: AFG Bioscience

Synonyms: Methyl 2,3,4-Tri-O-benzyl-α-D-glucopyranoside

C53239-1G C53239-2G C53239-5G
76619-860EA 155.76 CAD
76619-860 76619-862 76619-864
Methyl-2,3,4-tri-O-benzyl-ɑ-D-glucopyranoside ≥95%
Methyl-2,3,4-tri-O-benzyl-alpha-D-glucopyranoside
Formula: C₂₈H₃₂O₆
MW: 464.56 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06798958
CAS Number: 53008-65-4

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR