Methyl 2,3,4-tri-O-benzyl-α-D-galactopyranoside 95%

Supplier: AFG BIOSCIENCE LLC

C53215-1G C53215-2G C53215-5G
76619-854EA 194.7 CAD
76619-854 76619-856 76619-858
Methyl 2,3,4-tri-O-benzyl-α-D-galactopyranoside 95%
Methyl 2,3,4-tri-O-benzyl-α-D-galactopyranoside
Formula: C₂₈H₃₂O₆
MW: 464.55 g/mol
Storage Temperature: Freezer
CAS Number: 55094-38-7

Order Now

Specification Test Results

Purity Limit 95%
Appearance White solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR