Methyl 6-O-trityl-α-D-galactopyranoside 95%

Supplier: AFG Bioscience

C53214-10G C53214-1G C53214-5G
76619-852EA 778.81 CAD
76619-852 76619-848 76619-850
Methyl 6-O-trityl-α-D-galactopyranoside 95%
Methyl 6-O-trityl-α-D-galactopyranoside
Formula: C₂₆H₂₈O₆
MW: 436.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08274514
CAS Number: 35920-83-3

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR