α-Man-PEG3-Amine 95%

Supplier: AFG BIOSCIENCE LLC

C53010-25MG C53010-50MG
76619-844EA 395.6 CAD
76619-844 76619-846
α-Man-PEG3-Amine 95%
α-Man-PEG3-Amine
Formula: C₁₂H₂₅NO₈
MW: 311.33 g/mol
Storage Temperature: Freezer
CAS Number: 353737-46-9

Order Now

Specification Test Results

Purity Limit 95%
Appearance colorless sticky syrup


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR