β-Gal-PEG3-Amine 95%

Supplier: AFG Bioscience

C53008-25MG C53008-50MG
76619-836EA 395.6 CAD
76619-836 76619-838
β-Gal-PEG3-Amine 95%
β-Gal-PEG3-Amine
Formula: C₁₂H₂₅NO₈
Storage Temperature: Freezer
CAS Number: 126765-29-5

Order Now

Specification Test Results

Purity Limit 95%
Appearance colorless sticky syrup


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR