alpha-GalNAc-PEG3-Amine 95%

Supplier: AFG Bioscience

C53006-25MG C53006-50MG
76619-828EA 870.33 CAD
76619-828 76619-830
alpha-GalNAc-PEG3-Amine 95%
alpha-GalNAc-PEG3-Amine
Formula: C₁₄H₂₈N₂O₈
Storage Temperature: Refrigerator
CAS Number: 267227-84-9

Order Now

Specification Test Results

Purity Limit 95%
Appearance colorless sticky oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR