2-Deoxy-2-fluoro-L-fucose, 3,4-diacetate 95%

Supplier: AFG Bioscience

Synonyms: 3,4-Di-O-acetyl-2,6-dideoxy-2-fluoro-L-galactopyranose, 3,4-Di-O-acetyl-2-deoxy-2-fluoro-L-fucopyranose

C52517-100MG C52517-250MG C52517-50MG
76619-818EA 791.2 CAD
76619-818 76619-820 76619-816
2-Deoxy-2-fluoro-L-fucose, 3,4-diacetate 95%
2-Deoxy-2-fluoro-L-fucose, 3,4-diacetate
Formula: C₁₀H₁₅FO₆
MW: 250.22 g/mol
Storage Temperature: Freezer
CAS Number: 249566-79-8

Order Now

Specification Test Results

Purity Limit 95%
Appearance White solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR