6-Hexadecanoylamino-4-methylumbelliferyl β-D-Galactopyranoside ≥95%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
KRB, N-[7-(β-D-Galactopyranosyloxy)-4-methyl-2-oxo-2H-1-benzopyran-6-yl]- hexadecanamide, 6-HMU galactoside
C52306-10MG
C52306-2MG
C52306-5MG
76619-804EA
1342.57
CAD
76619-804
76619-800
76619-802
6-Hexadecanoylamino-4-methylumbelliferyl β-D-Galactopyranoside ≥95%
6-Hexadecanoylamino-4-methylumbelliferyl β-D-Galactopyranoside
Formula:
C₃₂H₄₉NO₉ MW: 591.73 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06796945 CAS Number: 94452-17-2 |
Specification Test Results
Purity Limit | > 95% |
Appearance | white powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...