6-NBDG ≥97%
Supplier: AFG Bioscience
Synonyms:
6-Deoxy-6-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose
C52304-10MG
C52304-5MG
C52304-1MG
76619-798EA
789.97
CAD
76619-798
76619-796
77888-862
6-NBDG ≥97%
6-NBDG
Formula:
C₁₂H₁₄N₄O₈ Storage Temperature: Freezer |
CAS Number:
108708-22-1 |
Specification Test Results
Purity Limit | > 97% |
Appearance | Crystalline Solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...