2-Propynyl-tetra-O-acetyl-β-D-glucopyranoside 97%

Supplier: AFG Bioscience

C52042-1G C52042-2G C52042-5G
76619-786EA 134.51 CAD
76619-786 76619-788 76619-790
2-Propynyl-tetra-O-acetyl-β-D-glucopyranoside 97%
2-Propynyl-tetra-O-acetyl-β-D-glucopyranoside
Formula: C₁₇H₂₂O₁₀
Storage Temperature: Refrigerator
MDL Number: MFCD03094034
CAS Number: 34272-02-1

Order Now

Specification Test Results

Purity Limit 97%
Appearance White Crystalline Solid

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR