2-​Deoxy-​2-​[(1-​oxo-​4-​pentyn-​1-​yl)​amino]​-α-​D-​glucopyranose 95%

Supplier: AFG Bioscience

C52020-100MG C52020-50MG
76619-772EA 1186.8 CAD
76619-772 76619-770
2-​Deoxy-​2-​[(1-​oxo-​4-​pentyn-​1-​yl)​amino]​-α-​D-​glucopyranose 95%
2-​Deoxy-​2-​[(1-​oxo-​4-​pentyn-​1-​yl)​amino]​-α-​D-​glucopyranose
Formula: C₁₁H₁₇NO₆
Storage Temperature: Freezer
CAS Number: 1635433-51-0

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR