2-propyn-1-yl 2-(acetylamino) β-D-Galactoside 95%

Supplier: AFG BIOSCIENCE LLC

C52019-100MG C52019-250MG
76619-766EA 553.84 CAD
76619-766 76619-768
2-propyn-1-yl 2-(acetylamino) β-D-Galactoside 95%
2-propyn-1-yl 2-(acetylamino) β-D-Galactoside
Formula: C₁₁H₁₇NO₆
MW: 259.26 g/mol
Storage Temperature: Freezer
CAS Number: 1611491-04-3

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR