2-Propynyl 2-acetamido-2-deoxy-α-D-glucopyranoside 95%

Supplier: AFG Bioscience

C52018-100MG C52018-250MG C52018-500MG
76619-760EA 311.53 CAD
76619-760 76619-762 76619-764
2-Propynyl 2-acetamido-2-deoxy-α-D-glucopyranoside 95%
2-Propynyl 2-acetamido-2-deoxy-α-D-glucopyranoside
Formula: C₁₁H₁₇NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD24386361
CAS Number: 1000593-95-2

Order Now

Specification Test Results

Purity Limit 95%
Appearance White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR