Propargyl β-D-lactoside 95%

Supplier: AFG BIOSCIENCE LLC

C52014-100MG C52014-250MG
76619-756EA 632.96 CAD
76619-756 76619-758
Propargyl β-D-lactoside 95%
Propargyl β-D-lactoside
Formula: C₁₅H₂₄O₁₁
MW: 380.34 g/mol
Storage Temperature: Freezer
CAS Number: 857642-16-1

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR