2-propyn-1-yl β-D-Glucoside 95%

Supplier: AFG Bioscience

C52011-100MG C52011-250MG
76619-748EA 316.47 CAD
76619-748 76619-750
2-propyn-1-yl β-D-Glucoside 95%
2-propyn-1-yl β-D-Glucoside
Formula: C₉H₁₄O₆
Storage Temperature: Freezer
CAS Number: 34272-03-2

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR