2-propyn-1-yl β-D-Galactoside 95%

Supplier: AFG BIOSCIENCE LLC

C52010-100MG C52010-250MG
76619-744EA 316.47 CAD
76619-744 76619-746
2-propyn-1-yl β-D-Galactoside 95%
2-propyn-1-yl β-D-Galactoside
Formula: C₉H₁₄O₆
MW: 218.2 g/mol
Storage Temperature: Freezer
CAS Number: 151168-59-1

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR