2-propyn-1-yl 2-(acetylamino) β-D-Glucoside 95%

Supplier: AFG BIOSCIENCE LLC

C52007-100MG C52007-250MG
76619-740EA 553.84 CAD
76619-740 76619-742
2-propyn-1-yl 2-(acetylamino) β-D-Glucoside 95%
2-propyn-1-yl 2-(acetylamino) β-D-Glucoside
Formula: C₁₁H₁₇NO₆
MW: 259.26 g/mol
Storage Temperature: Freezer
CAS Number: 390387-55-0

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR