β-Glc-PEG3-Alkyne 95%
Supplier: AFG Bioscience
C52004-25MG
C52004-50MG
C52004-100MG
C52004-10MG
76619-728EA
473.48
CAD
76619-728
76619-730
77888-829
77888-830
β-Glc-PEG3-Alkyne 95%
β-Glc-PEG3-Alkyne
Formula:
C₁₅H₂₆O₉ Storage Temperature: Freezer |
CAS Number:
1072903-76-4 |
Specification Test Results
Purity Limit | 95% |
Appearance | White powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...