β-Gal-PEG3-Alkyne 95%

Supplier: AFG Bioscience

C52003-25MG C52003-50MG C52003-100MG C52003-10MG
76619-724EA 473.48 CAD
76619-724 76619-726 77888-827 77888-828
β-Gal-PEG3-Alkyne 95%
β-Gal-PEG3-Alkyne
Formula: C₁₅H₂₆O₉
Storage Temperature: Freezer
CAS Number: 1072903-79-7

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR