β-GlcNAc-PEG3-Alkyne 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 2-{2-[2-(Prop-2-ynyloxy)ethoxy]ethoxy}ethyl 2-acetamido-2-deoxy-β-D-glucopyranoside, N-((2R,3R,4R,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-(2-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)ethoxy)tetrahydro-2H-pyran-3-yl)acetamide

C52002-25MG C52002-50MG
76619-720EA 467.29 CAD
76619-720 76619-722
β-GlcNAc-PEG3-Alkyne 95%
β-GlcNAc-PEG3-Alkyne
Formula: C₁₇H₂₉NO₉
MW: 391.41 g/mol
Storage Temperature: Freezer
CAS Number: 1884184-44-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR