3-Azido-3-deoxy-D-glucopyranose ≥95%

Supplier: AFG Bioscience

C51784-1G C51784-500MG C51784-250MG
76619-698EA 700.94 CAD
76619-698 76619-696 77888-818
3-Azido-3-deoxy-D-glucopyranose ≥95%
3-Azido-3-deoxy-D-glucopyranose
Formula: C₆H₁₁N₃O₅
Storage Temperature: Freezer
CAS Number: 104875-44-7

Order Now

Specification Test Results

Purity Limit > 95%
Appearance Colorless Low-Melting Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR