β-Lac-PEG3-Azide ≥95%

Supplier: AFG Bioscience

Synonyms: 2-[2-(2-Azidoethoxy)ethoxy]ethyl β-D-lactopyranoside

C51782-100MG C51782-250MG C51782-50MG
76619-688EA 1026.08 CAD
76619-688 76619-690 76619-686
β-Lac-PEG3-Azide ≥95%
β-Lac-PEG3-Azide
Formula: C₁₈H₃₃N₃O₁₃
MW: 499.47 g/mol
Storage Temperature: Freezer
CAS Number: 246855-74-3

Order Now

Specification Test Results

Purity Limit > 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR