2-azidoethyl α-L-Fucopyranoside ≥95%

Supplier: AFG Bioscience

Synonyms: 2-Azidoethyl 6-deoxy-α-L-galactopyranoside

C51780-100MG C51780-250MG C51780-500MG C51780-25MG
76619-680EA 631.73 CAD
76619-680 76619-682 76619-684 77888-815
2-azidoethyl α-L-Fucopyranoside ≥95%
2-azidoethyl α-L-Fucopyranoside
Formula: C₈H₁₅N₃O₅
MW: 233.22 g/mol
Storage Temperature: Refrigerator
CAS Number: 153252-86-9

Order Now

Specification Test Results

Purity Limit > 95%
Appearance Sticky Syrup


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR