3-Azidopropyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranoside ≥95%
Supplier: AFG Bioscience
C51610-1G
C51610-500MG
C51610-100MG
C51610-250MG
76619-634EA
1578.69
CAD
76619-634
76619-632
77888-784
77888-785
3-Azidopropyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranoside ≥95%
3-Azidopropyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranoside
Formula:
C₁₇H₂₆N₄O₉ MW: 430.41 g/mol Storage Temperature: Refrigerator |
CAS Number:
595568-98-2 |
Specification Test Results
Purity Limit | > 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...