2-​Acetamido-​2-​deoxy-​β-​D-​glucopyranosyl azide 95%

Supplier: AFG Bioscience

C51568-1G C51568-2G C51568-500MG
76619-590EA 389.41 CAD
76619-590 76619-592 76619-588
2-​Acetamido-​2-​deoxy-​β-​D-​glucopyranosyl azide 95%
2-​Acetamido-​2-​deoxy-​β-​D-​glucopyranosyl azide
Formula: C₈H₁₄N₄O₅
Storage Temperature: Freezer
MDL Number: MFCD00049240
CAS Number: 29847-23-2

Order Now

Specification Test Results

Purity Limit 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR